1,3-Diethynyl-5-(trifluoromethyl)benzene
Catalog # APOSPC50555-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:194.153 g/mol
Storage Temperature:Refrigerator
CAS Number:1622871-80-0
Formula:C₁₁H₅F₃