Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Diethynyl-5-fluorobenzene
1,3-Diethynyl-5-fluorobenzene 95%
1,3-Diethynyl-5-fluorobenzene 95%
1,3-Diethynyl-5-fluorobenzene 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1,3-Diethynyl-5-fluorobenzene 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
219633-74-6
:
Formula:
C₁₀H₅F
MDL Number:
MFCD18071615
:
144.145 g/mol
Storage Temperature:
Refrigerator