Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dichloro(trifluoromethoxy)benzene
3,5-Dichloro(trifluoromethoxy)benzene 98%
3,5-Dichloro(trifluoromethoxy)benzene 98%
3,5-Dichloro(trifluoromethoxy)benzene 98%
Supplier:
Apollo Scientific
Certificates
Expand image
3,5-Dichloro(trifluoromethoxy)benzene 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
151276-08-3
:
Formula:
C₇H₃Cl₂F₃O
MDL Number:
MFCD28787555
:
230.999 g/mol
Storage Temperature:
Ambient