Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Difluoro-3-(trifluoromethyl)bromobenzene
2,5-Difluoro-3-(trifluoromethyl)bromobenzene 97%
2,5-Difluoro-3-(trifluoromethyl)bromobenzene 97%
2,5-Difluoro-3-(trifluoromethyl)bromobenzene 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2,5-Difluoro-3-(trifluoromethyl)bromobenzene 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1807071-16-4
:
Formula:
C₇H₂BrF₅
MDL Number:
MFCD28737903
:
260.987 g/mol
Storage Temperature:
Ambient