Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dibromobenzyl fluoride
2,4-Dibromobenzyl fluoride 95%
2,4-Dibromobenzyl fluoride 95%
Corrosive
2,4-Dibromobenzyl fluoride 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
2,4-Dibromobenzyl fluoride 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1780254-42-3
:
Formula:
C₇H₅Br₂F
MDL Number:
MFCD00079786
:
267.921 g/mol
Storage Temperature:
Ambient
UN:
3265
ADR:
8,III
:
GHS05