Order Entry
Export
ContactUsLinkComponent
2,6-Difluoro-4-iodopyridine 97%
2,6-Difluoro-4-iodopyridine 97%
Catalog # APOSPC502363-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Difluoro-4-iodopyridine 97%
Catalog # APOSPC502363-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :240.977 g/mol
Storage Temperature:Ambient
CAS Number:685517-71-9
Formula:C₅H₂F₂IN
MDL Number:MFCD11977432