0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
3,4-Difluoro-2-(difluoromethoxy)nitrobenzene
3,4-Difluoro-2-(difluoromethoxy)nitrobenzene
3,4-Difluoro-2-(difluoromethoxy)nitrobenzene
Irritant
3,4-Difluoro-2-(difluoromethoxy)nitrobenzene
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,4-Difluoro-2-(difluoromethoxy)nitrobenzene
Supplier:
Apollo Scientific
About this item
CAS Number:
206546-18-1
:
Formula:
C₇H₃F₄NO₃
MDL Number:
MFCD22580776
:
225.097 g/mol
:
GHS07
Storage Temperature:
Ambient