Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Chloromethyl)-5-fluoro-1,3-benzoxazole
2-(Chloromethyl)-5-fluoro-1,3-benzoxazole
2-(Chloromethyl)-5-fluoro-1,3-benzoxazole
APOSPC502040-250MG
:
Apollo Scientific
:
2-(Chloromethyl)-5-fluoro-1,3-benzoxazole
APOSPC502040-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
185.583 g/mol
:
GHS07
:
Ambient
:
143708-35-4
:
C₈H₅ClFNO
:
MFCD10696613