Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-4,5-difluorobenzyl bromide
3-Bromo-4,5-difluorobenzyl bromide
3-Bromo-4,5-difluorobenzyl bromide
Corrosive
3-Bromo-4,5-difluorobenzyl bromide
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
3-Bromo-4,5-difluorobenzyl bromide
Supplier:
Apollo Scientific
About this item
CAS Number:
1807172-44-6
:
Formula:
C₇H₄Br₂F₂
MDL Number:
MFCD28737921
:
285.911 g/mol
Storage Temperature:
Ambient
UN:
3261
ADR:
8,III
:
GHS05