Order Entry
Export
ContactUsLinkComponent
3-Bromo-2,5-difluorobenzonitrile 98%
Toxic
3-Bromo-2,5-difluorobenzonitrile 98%
Catalog # APOSPC501939-250MG
Supplier:  Apollo Scientific
CAS Number:  
Toxic
3-Bromo-2,5-difluorobenzonitrile 98%
Catalog # APOSPC501939-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :217.998 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:1638487-41-8
Formula:C₇H₂BrF₂N
MDL Number:MFCD27923485