Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-bromo-2,5-difluorobenzoate
Methyl 4-bromo-2,5-difluorobenzoate
Methyl 4-bromo-2,5-difluorobenzoate
Irritant
Methyl 4-bromo-2,5-difluorobenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl 4-bromo-2,5-difluorobenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
1193162-21-8
:
Formula:
C₈H₅BrF₂O₂
MDL Number:
MFCD23713110
:
251.025 g/mol
:
GHS07
Storage Temperature:
Refrigerator