Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-2,5-difluorobenzaldehyde
3-Bromo-2,5-difluorobenzaldehyde 97%
3-Bromo-2,5-difluorobenzaldehyde 97%
Irritant
3-Bromo-2,5-difluorobenzaldehyde 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Bromo-2,5-difluorobenzaldehyde 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
112279-64-8
:
Formula:
C₇H₃BrF₂O
MDL Number:
MFCD26401549
:
220.999 g/mol
:
GHS07
Storage Temperature:
Ambient