4-Fluoro-5-methoxybenzene-1,2-diamine
Catalog # APOSPC501750-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:156.158 g/mol
Storage Temperature:Ambient
CAS Number:125163-15-7
Formula:C₇H₉FN₂O
MDL Number:MFCD18804706