6-Bromo-2,3-difluoroaniline 95+%
Catalog # APOSPC501691-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:208.003 g/mol
Storage Temperature:Ambient
CAS Number:887579-74-0
Formula:C₆H₄BrF₂N
MDL Number:MFCD03840796