Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Difluoro-4-iodo-1,3-benzodioxole
2,2-Difluoro-4-iodo-1,3-benzodioxole
2,2-Difluoro-4-iodo-1,3-benzodioxole
2,2-Difluoro-4-iodo-1,3-benzodioxole
Supplier:
Apollo Scientific
Certificates
Expand image
2,2-Difluoro-4-iodo-1,3-benzodioxole
Supplier:
Apollo Scientific
About this item
CAS Number:
531508-54-0
:
Formula:
C₇H₃F₂IO₂
MDL Number:
MFCD17016040
:
283.999 g/mol
Storage Temperature:
Ambient