Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Difluoro-4-iodobenzotrifluoride
2,6-Difluoro-4-iodobenzotrifluoride 98%
2,6-Difluoro-4-iodobenzotrifluoride 98%
:
Apollo Scientific
2,6-Difluoro-4-iodobenzotrifluoride 98%
:
Apollo Scientific
:
910787-71-2
:
:
C₇H₂F₅I
:
MFCD28396364
:
307.987 g/mol
:
GHS07
:
Ambient