2-Bromo-3,6-difluorophenol
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:1208077-18-2
- Formula:C₆H₃BrF₂O
- MDL Number:MFCD13176518
- Molecular Weight:208.988 g/mol
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
- CAS Number:1208077-18-2
- Formula:C₆H₃BrF₂O
- MDL Number:MFCD13176518
- Molecular Weight:208.988 g/mol
- Storage Temperature:Ambient



