0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
3-Chloro-4-fluorobenzene -1,2-diamine
3-Chloro-4-fluorobenzene -1,2-diamine 95%
3-Chloro-4-fluorobenzene -1,2-diamine 95%
3-Chloro-4-fluorobenzene -1,2-diamine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Chloro-4-fluorobenzene -1,2-diamine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
132915-81-2
:
Formula:
C₆H₆ClFN₂
MDL Number:
MFCD18804697
:
160.577 g/mol
Storage Temperature:
Refrigerator