Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-2,4-difluorobenzotrifluoride
5-Bromo-2,4-difluorobenzotrifluoride
5-Bromo-2,4-difluorobenzotrifluoride
:
Apollo Scientific
5-Bromo-2,4-difluorobenzotrifluoride
:
Apollo Scientific
:
261945-71-5
:
:
C₇H₂BrF₅
:
MFCD01631614
:
260.987 g/mol
:
GHS07
:
Ambient