

(3,4-Difluorophenyl)methanesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:163295-73-6
:
Formula:C₇H₅ClF₂O₂S
MDL Number:MFCD08704570
ADR:8,III
:226.628 g/mol
UN:3261
:GHS05
:GHS07
Storage Temperature:Ambient