Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromo-4,5-difluoro-1H-benzimidazole
6-Bromo-4,5-difluoro-1H-benzimidazole
6-Bromo-4,5-difluoro-1H-benzimidazole
APOSPC500001-1G
:
Apollo Scientific
:
6-Bromo-4,5-difluoro-1H-benzimidazole
APOSPC500001-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
233.013 g/mol
:
Ambient
:
1375068-79-3
:
C₇H₃BrF₂N₂
:
MFCD24369743