Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Difluoro-2-methylbenzaldehyde
3,6-Difluoro-2-methylbenzaldehyde 95%
3,6-Difluoro-2-methylbenzaldehyde 95%
:
Apollo Scientific
3,6-Difluoro-2-methylbenzaldehyde 95%
:
Apollo Scientific
:
1378525-21-3
:
:
C₈H₆F₂O
:
MFCD16250132
:
156.129 g/mol
:
GHS07
:
Ambient