0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
7-Fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
7-Fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
7-Fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
Irritant
7-Fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
7-Fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
886363-98-0
:
Formula:
C₈H₅FN₂O₂
MDL Number:
MFCD07374365
:
180.136 g/mol
:
GHS07
Storage Temperature:
Ambient