Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Nitro-6-bromo-8-fluoroimidazo[1,2-a]pyridine
3-Nitro-6-bromo-8-fluoroimidazo[1,2-a]pyridine
3-Nitro-6-bromo-8-fluoroimidazo[1,2-a]pyridine
Irritant
3-Nitro-6-bromo-8-fluoroimidazo[1,2-a]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Nitro-6-bromo-8-fluoroimidazo[1,2-a]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
2379918-59-7
:
Formula:
C₇H₃BrFN₃O₂
MDL Number:
MFCD31745720
:
260.02 g/mol
:
GHS07
Storage Temperature:
Ambient