Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
8-Fluoroimidazo[1,2-a]pyridine hydrate
8-Fluoroimidazo[1,2-a]pyridine hydrate
8-Fluoroimidazo[1,2-a]pyridine hydrate
Irritant
8-Fluoroimidazo[1,2-a]pyridine hydrate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
8-Fluoroimidazo[1,2-a]pyridine hydrate
Supplier:
Apollo Scientific
About this item
CAS Number:
2379918-38-2
:
Formula:
C₇H₅FN₂·H₂O
MDL Number:
MFCD31812097
:
154.142 g/mol
Melting Pt:
48…50 °C
:
GHS07
Storage Temperature:
Ambient