
2,6-Dibromo-4-[(trifluoromethyl)sulphonyl]phenol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:432-79-1
:
Formula:C₇H₃Br₂F₃O₃S
MDL Number:MFCD24369701
:383.965 g/mol
Melting Pt:150…154 °C
:GHS07
Storage Temperature:Ambient