Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-5-fluoro-1,3-thiazole hydrochloride
2-Amino-5-fluoro-1,3-thiazole hydrochloride 97%
2-Amino-5-fluoro-1,3-thiazole hydrochloride 97%
APOSPC49194-1G
:
Apollo Scientific
:
2-Amino-5-fluoro-1,3-thiazole hydrochloride 97%
APOSPC49194-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
154.594 g/mol
:
GHS07
:
Refrigerator
:
745053-64-9
:
C₃H₃FN₂S·ClH
:
MFCD09056854