

1,4-Bis(pentafluorothio)perfluorobenzene
Catalog # APOSPC49109-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:402.172 g/mol
Melting Pt:129…130 °C
Storage Temperature:Ambient
UN:1325
ADR:4.1,III
:GHS02
:GHS07
CAS Number:1219501-60-6
Formula:C₆F₁₄S₂
MDL Number:MFCD18071069