

2,4-Difluoro-3-(trifluoromethyl)benzyl bromide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1445995-80-1
:
Formula:C₈H₄BrF₅
MDL Number:MFCD25541882
:275.013 g/mol
UN:3265
ADR:8,II
:GHS05
:GHS07
Storage Temperature:Ambient