Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-2,4-difluoroaniline
3-Bromo-2,4-difluoroaniline
3-Bromo-2,4-difluoroaniline
:
Apollo Scientific
3-Bromo-2,4-difluoroaniline
:
Apollo Scientific
:
103977-79-3
:
:
C₆H₄BrF₂N
:
MFCD12407090
:
6.1,III
:
208.003 g/mol
:
2811
:
GHS06
:
Refrigerator