Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 2-chloro-4,4-difluoroacetoacetate
Ethyl 2-chloro-4,4-difluoroacetoacetate, tech
Ethyl 2-chloro-4,4-difluoroacetoacetate, tech
Irritant
Ethyl 2-chloro-4,4-difluoroacetoacetate, tech
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Ethyl 2-chloro-4,4-difluoroacetoacetate, tech
Supplier:
Apollo Scientific
About this item
CAS Number:
933772-60-2
:
Formula:
C₆H₇ClF₂O₃
MDL Number:
MFCD24477415
:
200.568 g/mol
:
GHS07
Storage Temperature:
Refrigerator