Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(1S)-2-Chloro-1-(2,4-difluorophenyl)ethanol
(1S)-2-Chloro-1-(2,4-difluorophenyl)ethanol 99%
(1S)-2-Chloro-1-(2,4-difluorophenyl)ethanol 99%
:
Apollo Scientific
(1S)-2-Chloro-1-(2,4-difluorophenyl)ethanol 99%
:
Apollo Scientific
:
330156-49-5
:
:
C₈H₇ClF₂O
:
MFCD09033383
:
192.59 g/mol
:
GHS05
:
GHS07
:
Ambient