Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dichloro-2-fluorobenzoic acid
3,5-Dichloro-2-fluorobenzoic acid
3,5-Dichloro-2-fluorobenzoic acid
Irritant
3,5-Dichloro-2-fluorobenzoic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,5-Dichloro-2-fluorobenzoic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
665022-07-1
:
Formula:
C₇H₃Cl₂FO₂
MDL Number:
MFCD04091131
:
209.002 g/mol
:
GHS07
Storage Temperature:
Ambient