

Ethyl 2,2-difluorohexanoate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:74106-81-3
:
Formula:C₈H₁₄F₂O₂
MDL Number:MFCD11007687
Boiling Pt:83…85 °C (15 mm Hg)
Storage Temperature:Ambient
UN:1993
ADR:3,III
:180.192 g/mol
:GHS02
:GHS07