Order Entry
Export
ContactUsLinkComponent
2,6-Dichloro-3,5-difluoro-4-(trifluoromethyl)phenylhydrazine
Irritant
2,6-Dichloro-3,5-difluoro-4-(trifluoromethyl)phenylhydrazine
Catalog # APOSPC4818-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,6-Dichloro-3,5-difluoro-4-(trifluoromethyl)phenylhydrazine
Catalog # APOSPC4818-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :281.01 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:111234-64-1
Formula:C₇H₃Cl₂F₅N₂
MDL Number:MFCD03424472