Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Difluoro-2-methoxyaniline
3,6-Difluoro-2-methoxyaniline 95%
3,6-Difluoro-2-methoxyaniline 95%
Toxic
3,6-Difluoro-2-methoxyaniline 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Toxic
3,6-Difluoro-2-methoxyaniline 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1261677-79-5
:
Formula:
C₇H₇F₂NO
MDL Number:
MFCD18394892
:
159.133 g/mol
Storage Temperature:
Ambient
UN:
2811
ADR:
6.1,III
:
GHS06