Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Difluoro-2-(trifluoromethyl)benzoic acid
3,6-Difluoro-2-(trifluoromethyl)benzoic acid 95%
3,6-Difluoro-2-(trifluoromethyl)benzoic acid 95%
Irritant
3,6-Difluoro-2-(trifluoromethyl)benzoic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,6-Difluoro-2-(trifluoromethyl)benzoic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
111595-72-3
:
Formula:
C₈H₃F₅O₂
MDL Number:
MFCD32066758
:
226.1 g/mol
:
GHS07
Storage Temperature:
Ambient