Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(2-Chloroethyl)-3-(3,4-difluorophenyl)urea
1-(2-Chloroethyl)-3-(3,4-difluorophenyl)urea
1-(2-Chloroethyl)-3-(3,4-difluorophenyl)urea
Irritant
1-(2-Chloroethyl)-3-(3,4-difluorophenyl)urea
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-(2-Chloroethyl)-3-(3,4-difluorophenyl)urea
Supplier:
Apollo Scientific
About this item
CAS Number:
2168354-72-9
:
Formula:
C₉H₉ClF₂N₂O
MDL Number:
MFCD31372392
:
234.63 g/mol
:
GHS07
Storage Temperature:
Ambient