Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dimethyl perfluoro-3,6-dioxaoctane-1,8-dioate
Dimethyl perfluoro-3,6-dioxaoctane-1,8-dioate 98%
Dimethyl perfluoro-3,6-dioxaoctane-1,8-dioate 98%
:
Apollo Scientific
Dimethyl perfluoro-3,6-dioxaoctane-1,8-dioate 98%
:
Apollo Scientific
:
24647-20-9
:
:
C₈H₆F₈O₆
:
MFCD03094248
:
350.117 g/mol
:
Ambient
Environmentally Preferable: