Order Entry
Export
ContactUsLinkComponent
(2-Bromo-1-ethoxy-2,2-difluoroethoxy)triethylsilane 98%
(2-Bromo-1-ethoxy-2,2-difluoroethoxy)triethylsilane 98%
Catalog # APOSPC450503-1G
Supplier:  Apollo Scientific
CAS Number:  
(2-Bromo-1-ethoxy-2,2-difluoroethoxy)triethylsilane 98%
Catalog # APOSPC450503-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :319.259 g/mol
Storage Temperature:Ambient
CAS Number:1922958-26-6
Formula:C₁₀H₂₁BrF₂O₂Si
MDL Number:MFCD28901345