2,2-Difluoroethyl 1H,1H-perfluorohexyl carbonate
Catalog # APOSPC450329-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:408.113 g/mol
Storage Temperature:Ambient
CAS Number:1980035-41-3
Formula:C₉H₅F₁₃O₃
MDL Number:MFCD28334322