2,2-Difluoroethyl 4,4,5,5,5-pentafluoropentyl carbonate
Catalog # APOSPC450326-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:286.144 g/mol
Storage Temperature:Ambient
CAS Number:1980040-02-5
Formula:C₈H₉F₇O₃
MDL Number:MFCD28334319