2,2-Difluoroethyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Catalog # APOSPC450312-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:290.108 g/mol
Storage Temperature:Ambient
CAS Number:1980076-06-9
Formula:C₇H₆F₈O₃
MDL Number:MFCD28334305