2,2-Difluoroethyl 2,2,3,3-tetrafluoropropyl carbonate
Catalog # APOSPC450300-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:240.1 g/mol
Storage Temperature:Ambient
CAS Number:1980045-84-8
Formula:C₆H₆F₆O₃
MDL Number:MFCD28334294