Order Entry
Export
ContactUsLinkComponent
2,2-Difluorobutane 97%
Flammable
Irritant
Compressed Gas
2,2-Difluorobutane 97%
Catalog # APOSPC450284-5G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
Compressed Gas
2,2-Difluorobutane 97%
Catalog # APOSPC450284-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:30…31 °C
Melting Pt:–114 °C
ADR:3,I
 :94.103 g/mol
UN:1993
 :GHS02
 :GHS07
 :GHS04
Storage Temperature:Ambient
CAS Number:353-81-1
Formula:C₄H₈F₂
MDL Number:MFCD20489351