Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium 2,2-difluorohexanoate
Sodium 2,2-difluorohexanoate
Sodium 2,2-difluorohexanoate
Sodium 2,2-difluorohexanoate
Supplier:
Apollo Scientific
Certificates
Expand image
Sodium 2,2-difluorohexanoate
Supplier:
Apollo Scientific
About this item
CAS Number:
1403667-43-5
:
Formula:
C₆H₉F₂O₂·Na
MDL Number:
MFCD23134591
:
174.121 g/mol
Storage Temperature:
Ambient