1,1,1,2,3,3-Hexafluoropropyl-2,2-difluoroethyl ether
Catalog # APOSPC450207-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.072 g/mol
Storage Temperature:Ambient
CAS Number:1309602-06-9
Formula:C₅H₄F₈O
MDL Number:MFCD19442560