
3,5-Difluorobenzaldehyde oxime
Catalog # APOSPC450197-10G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:157.118 g/mol
Melting Pt:95…96 °C
:GHS07
Storage Temperature:Ambient
CAS Number:677728-83-5
Formula:C₇H₅F₂NO
MDL Number:MFCD03412421