
Sodium 3,4-difluorophenylacetate
Catalog # APOSPC450193-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:194.111 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1980064-70-7
Formula:C₈H₅F₂O₂·Na
MDL Number:MFCD28054313