Perfluoro(1,2-diethylcyclohexane)
Catalog # APOSPC450121-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:500.075 g/mol
Storage Temperature:Ambient
CAS Number:75169-49-2
Formula:C₁₀F₂₀
MDL Number:MFCD18072165